Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003906.4 | NP_003897.2 | 11 | 2946 | ||
XM_005261203.4 | XP_005261260.1 | 12 | 3522 | |||
XM_005261204.4 | XP_005261261.1 | 12 | 3418 | |||
XM_005261205.3 | XP_005261262.1 | 12 | 3034 | |||
GenBank mRNA | AB011144.1 | - | 11 | 2516 | ||
AJ010089.1 | - | 11 | 2875 | |||
AK126313.1 | - | 2 | 1128 | |||
AK127239.1 | - | 1 | 835 | |||
BC104958.1 | - | 11 | 2914 | 8143961 | ||
BC104960.1 | - | 11 | 2914 | 8143963 |