Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034108.1 | NP_001029280.1 | 4 | 632 | ||
XM_006241126.2 | XP_006241188.1 | 4 | 747 | |||
XM_006241127.2 | XP_006241189.1 | 4 | 747 | |||
XM_008765201.1 | XP_008763423.1 | 4 | 747 | |||
GenBank mRNA | AB048838.1 | - | 3 | 596 | ||
AB048839.1 | - | 2 | 400 | |||
BC088286.1 | - | 3 | 591 | 7383057 | ||
FQ226318.1 | - | 3 | 453 |