Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034108.1 | NP_001029280.1 | 5 | 707 | ||
XM_006241126.2 | XP_006241188.1 | 5 | 872 | |||
XM_006241127.2 | XP_006241189.1 | 6 | 839 | |||
XM_008765201.1 | XP_008763423.1 | 5 | 836 | |||
GenBank mRNA | AB048838.1 | - | 4 | 671 | ||
AB048839.1 | - | 3 | 475 | |||
BC088286.1 | - | 4 | 666 | 7383057 | ||
FQ226318.1 | - | 4 | 578 |