Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034108.1 | NP_001029280.1 | 4, 5 | 665 | ||
GenBank mRNA | AB048838.1 | - | 3, 4 | 629 | ||
AB048839.1 | - | 2, 3 | 433 | |||
BC088286.1 | - | 3, 4 | 624 | 7383057 |