Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207614.1 | NP_997497.1 | 8 | 856 | ||
GenBank mRNA | AY572415.1 | - | 8 | 856 | ||
BC078924.1 | - | 8 | 909 | 7106266 |