Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020365.4 | NP_065098.1 | 11 | 1464 | ||
XM_017002747.1 | XP_016858236.1 | 12 | 1425 | |||
GenBank mRNA | AF257077.1 | - | 11 | 1327 | ||
AK022399.1 | - | 11 | 1326 | |||
AK024006.1 | - | 11 | 1352 | |||
AK314668.1 | - | 11 | 1337 | |||
BC021017.1 | - | 11 | 1315 | |||
CR457285.1 | - | 7 | 718 |