Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108747.1 | NP_001102217.1 | 4 | 649 | ||
XM_006241402.2 | XP_006241464.1 | 4 | 685 |