Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241463.2 | XP_006241525.1 | 8, 9 | 2646 | ||
XM_008765404.1 | XP_008763626.1 | 7, 8 | 2337 | |||
XM_008765405.1 | XP_008763627.1 | 8, 9 | 2641 | |||
XM_008765406.1 | XP_008763628.1 | 8, 9 | 2476 | |||
XM_008765407.1 | XP_008763629.1 | 7, 8 | 2337 |