Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198786.2 | NP_942081.2 | 2, 3 | 470 | ||
GenBank mRNA | AY079080.1 | - | 2, 3 | 330 | ||
AY079081.2 | - | 1, 2 | 583 | |||
BC062388.1 | - | 2, 3 | 470 | 5620891 |
Gene Symbol: LOC100911027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748700.3 | XP_003748748.1 | 2, 3 | 427 |