Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013219.1 | NP_001013237.1 | 3 | 673 | ||
XM_006241709.2 | XP_006241771.1 | 6 | 1645 | |||
XM_006241710.2 | XP_006241772.1 | 6 | 2563 | |||
XM_008765513.1 | XP_008763735.1 | 5 | 1590 | |||
GenBank mRNA | AJ748840.1 | - | 3 | 673 |