Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003909.4 | NP_003900.1 | 8, 9 | 797 | ||
XM_005251093.4 | XP_005251150.1 | 7, 8 | 749 | |||
XM_017013945.1 | XP_016869434.1 | 9, 10 | 922 | |||
GenBank mRNA | AB014536.1 | - | 8, 9 | 754 | ||
AF077226.2 | - | 7, 8 | 695 | |||
AK292912.1 | - | 8, 9 | 786 | |||
BC007017.1 | - | 8, 9 | 781 | |||
BC008385.1 | - | 8, 9 | 781 | |||
BC036242.1 | - | 8, 9 | 757 | 5300530 | ||
BC066597.1 | - | 8, 9 | 768 | 6168840 |