Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130566.1 | NP_001124038.1 | 10 | 1883 | ||
XM_008765580.1 | XP_008763802.1 | 9 | 4357 |