Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 15 | 1876 | ||
NM_153050.2 | NP_694690.1 | 15 | 1876 | |||
NM_153051.2 | NP_694691.1 | 15 | 1876 | |||
XM_005261803.2 | XP_005261860.1 | 16 | 1994 | |||
XM_005261804.3 | XP_005261861.1 | 15 | 1911 | |||
XM_005261805.2 | XP_005261862.1 | 16 | 1994 | |||
XM_005261806.2 | XP_005261863.1 | 15 | 1876 | |||
XM_005261808.2 | XP_005261865.1 | 16 | 1994 | |||
XM_017029028.1 | XP_016884517.1 | 14 | 1768 | |||
XM_017029029.1 | XP_016884518.1 | 14 | 1758 | |||
XM_017029030.1 | XP_016884519.1 | 14 | 1774 | |||
XM_017029031.1 | XP_016884520.1 | 14 | 1762 | |||
XM_017029032.1 | XP_016884521.1 | 15 | 1759 | |||
XM_017029033.1 | XP_016884522.1 | 9 | 1195 | |||
XM_017029034.1 | XP_016884523.1 | 16 | 1970 | |||
GenBank mRNA | AB002369.1 | - | 15 | 1795 | ||
AF233436.2 | - | 12 | 1557 | |||
AF233437.1 | - | 12 | 1557 | |||
AF233438.1 | - | 12 | 1557 | |||
AK289581.1 | - | 15 | 1876 | |||
BC142713.1 | - | 15 | 1832 | 40148839 | ||
BC148216.1 | - | 14 | 1695 | 40082266 | ||
BC151217.1 | - | 15 | 1890 | 40148098 | ||
BC152455.1 | - | 15 | 1795 | 8862588 | ||
CR456525.1 | - | 13 | 1572 |