Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 8 | 836 | ||
NM_153050.2 | NP_694690.1 | 8 | 836 | |||
NM_153051.2 | NP_694691.1 | 8 | 836 | |||
XM_005261803.2 | XP_005261860.1 | 9 | 954 | |||
XM_005261804.3 | XP_005261861.1 | 8 | 871 | |||
XM_005261805.2 | XP_005261862.1 | 9 | 954 | |||
XM_005261806.2 | XP_005261863.1 | 8 | 836 | |||
XM_005261808.2 | XP_005261865.1 | 9 | 954 | |||
XM_017029028.1 | XP_016884517.1 | 7 | 728 | |||
XM_017029029.1 | XP_016884518.1 | 7 | 718 | |||
XM_017029030.1 | XP_016884519.1 | 7 | 734 | |||
XM_017029031.1 | XP_016884520.1 | 7 | 722 | |||
XM_017029032.1 | XP_016884521.1 | 8 | 719 | |||
XM_017029033.1 | XP_016884522.1 | 2 | 155 | |||
XM_017029034.1 | XP_016884523.1 | 8 | 809 | |||
GenBank mRNA | AB002369.1 | - | 8 | 755 | ||
AF233436.2 | - | 5 | 517 | |||
AF233437.1 | - | 5 | 517 | |||
AF233438.1 | - | 5 | 517 | |||
AK289581.1 | - | 8 | 836 | |||
BC142713.1 | - | 8 | 792 | 40148839 | ||
BC148216.1 | - | 7 | 655 | 40082266 | ||
BC151217.1 | - | 8 | 850 | 40148098 | ||
BC152455.1 | - | 8 | 755 | 8862588 | ||
CR456525.1 | - | 6 | 532 |