Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034940.1 | NP_001030112.1 | 3, 4 | 247 | ||
GenBank mRNA | BC099747.1 | - | 3, 4 | 247 | 5598773 |