Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134585.2 | NP_001128057.1 | 6 | 1877 | ||
XM_006242645.2 | XP_006242707.1 | 6 | 2602 | |||
XM_006242647.2 | XP_006242709.1 | 7 | 2204 | |||
XM_006242648.2 | XP_006242710.1 | 7 | 2778 | |||
XM_008765958.1 | XP_008764180.1 | 5 | 3022 |