Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272071.1 | NP_001259000.1 | 2 | 431 | ||
NM_003916.4 | NP_003907.3 | 2 | 431 | |||
XM_005274614.3 | XP_005274671.1 | 2 | 383 | |||
XM_011545599.1 | XP_011543901.1 | 2 | 383 | |||
XM_017029925.1 | XP_016885414.1 | 2 | 383 | |||
XM_017029926.1 | XP_016885415.1 | 2 | 318 | |||
GenBank mRNA | AB015320.1 | - | 1 | 104 | ||
AF087876.1 | - | 1 | 222 | |||
AF091077.1 | - | 1 | 105 | |||
AF251295.1 | - | 2 | 184 | |||
AK130075.1 | - | 2 | 233 | |||
AK296135.1 | - | 2 | 230 | |||
AK299921.1 | - | 2 | 230 | |||
AK302908.1 | - | 2 | 201 | |||
AK312594.1 | - | 2 | 230 | |||
BC001117.2 | - | 2 | 178 | 3050001 | ||
BC071867.1 | - | 2 | 180 | 4556064 | ||
BI462192.1 | - | 2 | 396 | |||
BT006738.1 | - | 1 | 104 | |||
BX537780.1 | - | 1 | 32 |
Gene Symbol: LOC653653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027408.1 | 1 | 361 | |||
GenBank mRNA | AK295685.1 | - | 1 | 361 |