Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198788.2 | NP_942083.1 | 1, 2 | 66 | ||
GenBank mRNA | BC059160.1 | - | 1, 2 | 71 | 6887444 | |
FQ209438.1 | - | 1, 2 | 67 | |||
FQ212791.1 | - | 1, 2 | 69 | |||
FQ212873.1 | - | 1, 2 | 69 | |||
FQ212985.1 | - | 1, 2 | 68 | |||
FQ213509.1 | - | 1, 2 | 68 | |||
FQ214315.1 | - | 1, 2 | 96 | |||
FQ215255.1 | - | 1, 2 | 73 | |||
FQ215355.1 | - | 1, 2 | 67 | |||
FQ215505.1 | - | 1, 2 | 68 | |||
FQ215951.1 | - | 1, 2 | 68 | |||
FQ215986.1 | - | 1, 2 | 67 | |||
FQ216027.1 | - | 1, 2 | 68 | |||
FQ216314.1 | - | 1, 2 | 74 | |||
FQ216503.1 | - | 1, 2 | 71 | |||
FQ220862.1 | - | 1, 2 | 67 | |||
FQ230166.1 | - | 1, 2 | 60 |