Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029924.1 | NP_001025095.1 | 8 | 770 | ||
GenBank mRNA | BC097373.1 | - | 8 | 770 | 7444462 |