Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl24d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014212.1 | NP_001014234.1 | 3, 4 | 321 | ||
GenBank mRNA | BC062237.1 | - | 3, 4 | 321 | 6919069 |