Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108775.1 | NP_001102245.1 | 2 | 79 | ||
XM_006243669.2 | XP_006243731.1 | 3 | 383 |