Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108777.1 | NP_001102247.1 | 3 | 1270 | ||
XM_006243800.1 | XP_006243862.1 | 3 | 1297 | |||
XM_006243801.2 | XP_006243863.1 | 3 | 948 | |||
GenBank mRNA | BC168984.1 | - | 3 | 1242 | 7103631 |