Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108778.1 | NP_001102248.1 | 12 | 1215 | ||
GenBank mRNA | BC158778.1 | - | 12 | 1212 | 7134304 | |
FQ226641.1 | - | 8 | 677 | |||
FQ230405.1 | - | 12 | 1198 |