Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110838.1 | NP_001104308.1 | 11 | 1471 | ||
XM_006244192.2 | XP_006244254.1 | 11 | 1589 | |||
XM_006244193.2 | XP_006244255.1 | 11 | 1698 | |||
XM_006244194.2 | XP_006244256.1 | 11 | 1614 | |||
XM_006244195.2 | XP_006244257.1 | 11 | 1582 | |||
XM_008766717.1 | XP_008764939.1 | 11 | 1614 | |||
XM_008766718.1 | XP_008764940.1 | 10 | 1723 | |||
XM_008766719.1 | XP_008764941.1 | 10 | 1481 | |||
XM_008766720.1 | XP_008764942.1 | 10 | 1446 | |||
XM_008766721.1 | XP_008764943.1 | 10 | 1478 | |||
XM_008766723.1 | XP_008764945.1 | 10 | 1593 | |||
XM_008766724.1 | XP_008764946.1 | 11 | 1506 | |||
XM_008766725.1 | XP_008764947.1 | 11 | 1510 | |||
XM_008766726.1 | XP_008764948.1 | 10 | 1374 | |||
GenBank mRNA | FQ210480.1 | - | 11 | 1491 | ||
FQ211645.1 | - | 11 | 1562 | |||
FQ215070.1 | - | 11 | 1491 |