Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108789.3 | NP_001102259.2 | 20 | 2345 | ||
GenBank mRNA | DN935088.1 | - | 3 | 309 |