Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108791.1 | NP_001102261.1 | 12, 13 | 4017 | ||
XM_006244673.2 | XP_006244735.1 | 11, 12 | 3703 | |||
XM_006244675.2 | XP_006244737.1 | 11, 12 | 3870 | |||
XM_006244676.2 | XP_006244738.1 | 11, 12 | 3763 | |||
XM_006244677.2 | XP_006244739.1 | 12, 13 | 3687 | |||
XM_008766954.1 | XP_008765176.1 | 12, 13 | 3640 |