Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128079.1 | NP_001121551.1 | 5, 6 | 501 | ||
XM_006245236.2 | XP_006245298.1 | 4, 5 | 1274 | |||
GenBank mRNA | BC161976.1 | - | 5, 6 | 501 | 9029838 |