Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128079.1 | NP_001121551.1 | 8 | 793 | ||
XM_006245236.2 | XP_006245298.1 | 7 | 1566 | |||
GenBank mRNA | BC105903.1 | - | 3 | 288 | ||
BC161976.1 | - | 8 | 793 | 9029838 |