Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025745.1 | NP_001020916.1 | 3, 4 | 488 | ||
XM_006245245.2 | XP_006245307.1 | 4, 5 | 394 | |||
XM_006245246.2 | XP_006245308.1 | 4, 5 | 376 | |||
GenBank mRNA | BC099215.1 | - | 3, 4 | 488 | 7384656 |