Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 27 | 5580 | ||
XM_006245252.2 | XP_006245314.1 | 29 | 5542 | |||
XM_006245255.2 | XP_006245317.1 | 22 | 5338 | |||
XM_006245256.2 | XP_006245318.1 | 22 | 3367 | |||
XM_008767267.1 | XP_008765489.1 | 27 | 5642 | |||
XM_008767268.1 | XP_008765490.1 | 27 | 5419 | |||
XM_008767269.1 | XP_008765491.1 | 28 | 6036 |
Set Membership: |
G-protein coupled receptor |