Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 21 | 4947 | ||
XM_006245252.2 | XP_006245314.1 | 23 | 4909 | |||
XM_006245255.2 | XP_006245317.1 | 16 | 4705 | |||
XM_006245256.2 | XP_006245318.1 | 16 | 2734 | |||
XM_008767267.1 | XP_008765489.1 | 21 | 5009 | |||
XM_008767268.1 | XP_008765490.1 | 21 | 4786 | |||
XM_008767269.1 | XP_008765491.1 | 22 | 5403 |
Set Membership: |
G-protein coupled receptor |