Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108809.3 | NP_001102279.2 | 5 | 532 | ||
XM_006245419.2 | XP_006245481.1 | 5 | 710 | |||
XM_006245420.2 | XP_006245482.1 | 5 | 709 |