Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007755.1 | NP_001007756.1 | 5 | 493 | ||
XM_006245431.2 | XP_006245493.1 | 5 | 1069 | |||
XM_008767276.1 | XP_008765498.1 | 5 | 1097 | |||
XM_008767277.1 | XP_008765499.1 | 5 | 572 | |||
GenBank mRNA | BC079358.1 | - | 5 | 493 | 7108615 |