Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037209.1 | NP_001032286.1 | 3 | 585 | ||
GenBank mRNA | BC098676.1 | - | 3 | 585 | 7372734 |