Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135607.1 | NP_001129079.1 | 435 | |||
XM_008773382.1 | XP_008771604.1 | 236 | ||||
XM_008773383.1 | XP_008771605.1 | 3 | 182 | |||
XM_008773384.1 | XP_008771606.1 | 121 | ||||
GenBank mRNA | DQ058651.1 | - | 435 |
Gene Symbol: LOC103690001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257477.2 | XP_006257539.1 | 3 | 182 | ||
XM_006257478.2 | XP_006257540.1 | 2 | 121 | |||
XM_008773510.1 | XP_008771732.1 | 3 | 236 |