Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276270.1 | NP_001263199.1 | 5 | 1710 | ||
NM_001276271.1 | NP_001263200.1 | 5 | 1728 | |||
NM_001276272.1 | NP_001263201.1 | 5 | 1728 | |||
NM_001276273.1 | NP_001263202.1 | 4 | 774 | |||
NM_003925.2 | NP_003916.1 | 5 | 1728 | |||
XM_011513268.2 | XP_011511570.1 | 4 | 522 | |||
GenBank mRNA | AF072250.1 | - | 5 | 1558 | ||
AF114784.1 | - | 5 | 1523 | |||
AF532602.1 | - | 5 | 1556 | |||
AK303013.1 | - | 5 | 1584 | |||
AK312878.1 | - | 5 | 1466 | |||
AM180876.1 | - | 4 | 428 | |||
BC011752.2 | - | 5 | 1407 | 3534047 | ||
BC034463.1 | - | 5 | 1382 | |||
CR450305.1 | - | 5 | 1364 |