Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256749.2 | XP_006256811.1 | 11 | 1299 | ||
XM_006256750.2 | XP_006256812.1 | 11 | 1299 | |||
XM_006256753.2 | XP_006256815.1 | 11 | 1141 | |||
XM_008773086.1 | XP_008771308.1 | 11 | 1520 | |||
XM_008773087.1 | XP_008771309.1 | 11 | 1296 | |||
XM_008773088.1 | XP_008771310.1 | 11 | 923 | |||
XM_008773090.1 | XP_008771312.1 | 11 | 1140 | |||
XM_008773091.1 | XP_008771313.1 | 11 | 1263 | |||
XM_008773092.1 | XP_008771314.1 | 7 | 2213 |