Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134588.1 | NP_001128060.1 | 5 | 420 | ||
XM_006256865.2 | XP_006256927.1 | 5 | 475 | |||
XM_006256866.2 | XP_006256928.1 | 5 | 474 | |||
XM_008773174.1 | XP_008771396.1 | 5 | 474 |