Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773209.1 | XP_008771431.1 | 3 | 583 | ||
XM_008773210.1 | XP_008771432.1 | 3 | 583 |