Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135662.1 | NP_001129134.1 | 5 | 661 | ||
NM_001135663.1 | NP_001129135.1 | 3 | 876 | |||
NM_001135664.1 | NP_001129136.1 | 4 | 508 | |||
NM_003929.2 | NP_003920.1 | 5 | 762 | |||
XM_005245569.1 | XP_005245626.1 | 5 | 660 | |||
XM_005245570.1 | XP_005245627.1 | 5 | 713 | |||
XM_005245571.1 | XP_005245628.1 | 5 | 638 | |||
XM_006711605.3 | XP_006711668.1 | 4 | 404 | |||
XM_006711606.2 | XP_006711669.1 | 4 | 368 | |||
XM_017002748.1 | XP_016858237.1 | 4 | 663 | |||
XM_017002749.1 | XP_016858238.1 | 4 | 641 | |||
XM_017002750.1 | XP_016858239.1 | 4 | 573 | |||
GenBank mRNA | AK223514.1 | - | 4 | 538 | ||
AK303879.1 | - | 3 | 627 | |||
AK308359.1 | - | 4 | 477 | |||
AK311085.1 | - | 3 | 385 | |||
AK313000.1 | - | 5 | 650 | |||
BC002585.2 | - | 5 | 595 | 3140744 | ||
BT019540.1 | - | 4 | 422 | |||
BX647960.1 | - | 5 | 727 | |||
CR542010.1 | - | 4 | 422 | |||
D84488.1 | - | 4 | 462 | |||
DA377295.1 | - | 4 | 463 |