Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135662.1 | NP_001129134.1 | 5 | 685 | ||
NM_001135663.1 | NP_001129135.1 | 3 | 900 | |||
NM_001135664.1 | NP_001129136.1 | 4 | 532 | |||
NM_003929.2 | NP_003920.1 | 5 | 786 | |||
XM_005245569.1 | XP_005245626.1 | 5 | 684 | |||
XM_005245570.1 | XP_005245627.1 | 5 | 737 | |||
XM_005245571.1 | XP_005245628.1 | 5 | 662 | |||
XM_006711605.3 | XP_006711668.1 | 4 | 428 | |||
XM_006711606.2 | XP_006711669.1 | 4 | 392 | |||
XM_017002748.1 | XP_016858237.1 | 4 | 687 | |||
XM_017002749.1 | XP_016858238.1 | 4 | 665 | |||
XM_017002750.1 | XP_016858239.1 | 4 | 597 | |||
GenBank mRNA | AK223514.1 | - | 4 | 562 | ||
AK303879.1 | - | 3 | 651 | |||
AK308359.1 | - | 4 | 501 | |||
AK311085.1 | - | 3 | 409 | |||
AK313000.1 | - | 5 | 674 | |||
BC002585.2 | - | 5 | 619 | 3140744 | ||
BT019540.1 | - | 4 | 446 | |||
BX647960.1 | - | 5 | 751 | |||
CR542010.1 | - | 4 | 446 | |||
D84488.1 | - | 4 | 486 | |||
DA377295.1 | - | 4 | 487 |