Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282112.1 | NP_001269041.1 | 10 | 1246 | ||
NM_001282113.1 | NP_001269042.1 | 10 | 1239 | |||
NM_003935.4 | NP_003926.1 | 10 | 1562 | |||
XM_005261811.1 | XP_005261868.1 | 12 | 1412 | |||
XM_005261813.2 | XP_005261870.1 | 11 | 1277 | |||
XM_006724349.1 | XP_006724412.1 | 11 | 1223 | |||
XM_006724350.1 | XP_006724413.1 | 11 | 1397 | |||
XM_011530482.1 | XP_011528784.1 | 11 | 1479 | |||
XM_017029038.1 | XP_016884527.1 | 10 | 1225 | |||
XM_017029039.1 | XP_016884528.1 | 12 | 1287 | |||
XM_017029040.1 | XP_016884529.1 | 11 | 1263 | |||
XM_017029041.1 | XP_016884530.1 | 12 | 1305 | |||
GenBank mRNA | AF017146.1 | - | 9 | 1147 | ||
AF053082.1 | - | 9 | 1147 | |||
AF125216.1 | - | 9 | 1107 | |||
AK096695.1 | - | 3 | 230 | |||
AK290997.1 | - | 10 | 1192 | |||
AK304709.1 | - | 8 | 897 | |||
AL833505.1 | - | 11 | 1276 | |||
BC002432.2 | - | 10 | 1469 | 3346377 | ||
CR456596.1 | - | 9 | 1061 |