Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013229.1 | NP_001013247.1 | 5, 6 | 594 | ||
XM_008768087.1 | XP_008766309.1 | 5, 6 | 590 | |||
GenBank mRNA | BC089885.1 | - | 5, 6 | 594 | 7370040 |