Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246915.2 | XP_006246977.1 | 15 | 3212 | ||
XM_006246917.2 | XP_006246979.1 | 13 | 2995 | |||
XM_008768139.1 | XP_008766361.1 | 11 | 2419 |