Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KYNU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032998.1 | NP_001028170.1 | 2, 3 | 307 | ||
NM_001199241.1 | NP_001186170.1 | 3, 4 | 393 | |||
NM_003937.2 | NP_003928.1 | 2, 3 | 307 | |||
XM_017005217.1 | XP_016860706.1 | 2, 3 | 269 | |||
GenBank mRNA | AK223426.1 | - | 2, 3 | 305 | ||
AK290900.1 | - | 2, 3 | 253 | |||
AK292095.1 | - | 2, 3 | 271 | |||
AK315343.1 | - | 3, 4 | 355 | |||
AL548658.3 | - | 2, 3 | 435 | |||
BC000879.1 | - | 2, 3 | 276 | 3461828 | ||
CB151988.1 | - | 2, 3 | 307 | |||
CR457423.1 | - | 1, 2 | 177 | |||
U57721.1 | - | 2, 3 | 283 |