Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261826.1 | NP_001248755.1 | 21 | 2627 | ||
NM_003938.6 | NP_003929.4 | 21 | 2627 | |||
XM_006722932.2 | XP_006722995.1 | 21 | 2690 | |||
XM_017027422.1 | XP_016882911.1 | 16 | 1924 | |||
GenBank mRNA | AB208804.1 | - | 21 | 2611 | ||
AF002163.1 | - | 2715 | ||||
BC005142.2 | - | 21 | 2455 | |||
BC022837.1 | - | 6 | 666 | |||
U91930.1 | - | 21 | 2340 |