Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108848.1 | NP_001102318.1 | 2 | 216 | ||
GenBank mRNA | AB212679.1 | - | 1 | 64 | ||
BC168972.1 | - | 2 | 222 | 9034405 |