Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108850.2 | NP_001102320.1 | 2 | 584 | ||
XM_006248770.2 | XP_006248832.1 | 2 | 1878 | |||
GenBank mRNA | BC158788.1 | - | 2 | 584 | 7305395 | |
FQ227574.1 | - | 2 | 599 |