Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015030.1 | NP_001015030.1 | 4 | 1041 | ||
GenBank mRNA | BC091342.1 | - | 4 | 1041 | 7315459 |