Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108853.1 | NP_001102323.1 | 9 | 962 | ||
XM_008769200.1 | XP_008767422.1 | 8 | 1020 |